Learn More
Invitrogen™ RP1 Polyclonal Antibody

Description
Immunogen sequence: VTCSPCEMCT VNKAYSPKET CNPSDTFFPS DGYGVDQTSM NKACFLGEVC SLTDTVFSDK ACAQKENHTY EGACPIDETY VPVNVCNTID FLNSKENTYT DNLDSTEELE RGDDIQKDLN ILTDPEYKNG FNTLVSHQNV SNLSSCG Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 49%.
Specifications
Specifications
| Antigen | RP1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RP1 |
| Gene Accession No. | P56715 |
| Gene Alias | Dcdc3; DCDC4A; mG145; Orp1; oxygen-regulated protein 1; retinitis pigmentosa 1 (autosomal dominant); retinitis pigmentosa 1 (human); retinitis pigmentosa 1 homolog; retinitis pigmentosa 1 protein; Retinitis pigmentosa RP1 protein; retinitis pigmentosa RP1 protein homolog; RP1; Rp1h |
| Gene Symbols | RP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.