Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572993
Description
Predicted to react with monkey based upon 100% sequence homology. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.
Microtubule-associated protein regulating the stability and length of the microtubule-based axoneme of photoreceptors. Required for the differentiation of photoreceptor cells, it plays a role in the organization of the outer segment of rod and cone photoreceptors ensuring the correct orientation and higher-order stacking of outer segment disks along the photoreceptor axoneme.
Specifications
| RP1 | |
| Polyclonal | |
| Unconjugated | |
| RP1 | |
| Dcdc3; DCDC4A; mG145; Orp1; oxygen-regulated protein 1; retinitis pigmentosa 1 (autosomal dominant); retinitis pigmentosa 1 (human); retinitis pigmentosa 1 homolog; retinitis pigmentosa 1 protein; Retinitis pigmentosa RP1 protein; retinitis pigmentosa RP1 protein homolog; RP1; Rp1h | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19888, 6101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 0.05% sodium azide | |
| P56715, P56716 | |
| RP1 | |
| A synthetic peptide made to a C-terminal portion of the human RP1 protein (between residues 2000-2156). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction