Learn More
RP105/CD180 Antibody (105), Alexa Fluor™ 750, Novus Biologicals™
SDP
Rabbit Monoclonal Antibody
Supplier:  Novus Biologicals NBP290611AF750
Catalog No. NB040238
Only null left
Add to Cart
Description
Description
RP105/CD180 Monoclonal specifically detects RP105/CD180 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
RP105/CD180
Monoclonal
Alexa Fluor 750
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
105
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse RP105/CD180 (NP_032559.2; Met1-Ser626).
0.1 mL
Adaptive Immunity, Immunology, Innate Immunity
4064
Store at 4°C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.