Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RP105/CD180 Antibody (G28-8), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262228JF646
Description
RP105/CD180 Monoclonal specifically detects RP105/CD180 in Human samples. It is validated for Flow Cytometry.Specifications
RP105/CD180 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD180 | |
Human tonsillar B cells | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
G28-8 | |
Flow Cytometry | |
CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology, Innate Immunity | |
4064 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction