Learn More
Invitrogen™ RP2 Monoclonal Antibody (3D7)

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 tissue, human Jurkat, whole cell, human Caco-2 whole cell, human Hela whole cell, human A549 whole cell. ICC/IF: U2OS cell. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Antigen | RP2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 3D7 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | RP2 |
| Gene Accession No. | O75695, Q9EPK2 |
| Gene Alias | AI662636; DELXp11.3; KIAA0215; NM23-H10; NME10; Protein XRP2; retinitis pigmentosa 2 (X-linked recessive); retinitis pigmentosa 2 homolog (human); RGD1565124; Rp2; RP2 activator of ARL3 GTPase; RP2, ARL3 GTPase activating protein; Rp2h; TBCCD2; XRP2; XRP2 protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.