Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564873 Supplier Invitrogen™ Supplier No. PA564873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELNWSLLPEDA VVQDYVPIPT TEELKAVRVS TEANRSIVPI SRGQRQKSSD ESCLVVLFAG DYTIANARKL IDEMVGKGFF LVQTKEVSMK AEDAQRVFRE KAPDFLPLLN KGPVIALE Highest antigen sequence identity to the following orthologs - mouse 88%, rat 87%.

The RP2 locus has been implicated as one cause of X-linked retinitis pigmentosa. The predicted gene product shows homology with human cofactor C, a protein involved in the ultimate step of beta-tubulin folding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RP2
Gene Accession No. O75695
Gene Alias AI662636; DELXp11.3; KIAA0215; NM23-H10; NME10; Protein XRP2; retinitis pigmentosa 2 (X-linked recessive); retinitis pigmentosa 2 homolog (human); RGD1565124; Rp2; RP2 activator of ARL3 GTPase; RP2, ARL3 GTPase activating protein; Rp2h; TBCCD2; XRP2; XRP2 protein
Gene Symbols RP2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human RP2 (Product #RP-91928)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6102
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.