Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RP5-1077B9.4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | RP5-1077B9.4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
RP5-1077B9.4 Polyclonal specifically detects RP5-1077B9.4 in Human, Mouse samples. It is validated for Western Blot.Specifications
RP5-1077B9.4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
60672 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
FLJ12438, FLJ38609, IGFBP2-binding protein, IGFBP-2-Binding Protein, IIp45, invasion inhibitory protein 45, Invasion-inhibitory protein 45, migration and invasion inhibitory protein, migration and invasion-inhibitory protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 279-388 of human RP5-1077B9.4 (NP_068752.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title