Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556769
Description
Immunogen sequence: PEQELQRRRE QKRRRHDAQQ LQQLKHLESF YEKPPPGLIK EDETKPEDCI PDVPGNEHAR EFLAHAPTKG LWM Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 90%.
Is thought to be a target protein for the PIM1 kinase. May play some roles in B-cell proliferation in association with PIM1.
Specifications
RP9 | |
Polyclonal | |
Unconjugated | |
RP9 | |
LOW QUALITY PROTEIN: retinitis pigmentosa 9 protein; PAP1; PAP-1; Pim-1 associated protein; Pim-1 kinase associated protein; pim-1-associated protein; retinitis pigmentosa 9 (autosomal dominant); retinitis pigmentosa 9 (human); retinitis pigmentosa 9 homolog; retinitis pigmentosa 9 protein; Retinitis pigmentosa 9 protein homolog; RGD1559759; RP9; RP9 pre-mRNA splicing factor; Rp9h | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TA86 | |
RP9 | |
Recombinant protein corresponding to Human RP9. Recombinant protein control fragment (Product #RP-92790). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction