Learn More
Invitrogen™ RP9 Polyclonal Antibody

Description
Immunogen sequence: GKEVKVMQCWR CKRYGHRTGD KECPFFIKGN QKLEQFRVAH EDPMYDIIRD NKRHEKDVRI QQLKQLLEDS TSD Highest antigen sequence identity to the following orthologs - mouse 97%, rat 80%.
Specifications
Specifications
| Antigen | RP9 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RP9 |
| Gene Accession No. | Q8TA86 |
| Gene Alias | LOW QUALITY PROTEIN: retinitis pigmentosa 9 protein; PAP1; PAP-1; Pim-1 associated protein; Pim-1 kinase associated protein; pim-1-associated protein; retinitis pigmentosa 9 (autosomal dominant); retinitis pigmentosa 9 (human); retinitis pigmentosa 9 homolog; retinitis pigmentosa 9 protein; Retinitis pigmentosa 9 protein homolog; RGD1559759; RP9; RP9 pre-mRNA splicing factor; Rp9h |
| Gene Symbols | RP9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.