Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570635 Supplier Invitrogen™ Supplier No. PA570635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Dog: 86%; Horse: 86%; Human: 100%; Mouse: 79%; Pig: 93%; Rat: 79%.

Is thought to be a target protein for the PIM1 kinase. May play some roles in B-cell proliferation in association with PIM1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RP9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RP9
Gene Accession No. Q8TA86
Gene Alias LOW QUALITY PROTEIN: retinitis pigmentosa 9 protein; PAP1; PAP-1; Pim-1 associated protein; Pim-1 kinase associated protein; pim-1-associated protein; retinitis pigmentosa 9 (autosomal dominant); retinitis pigmentosa 9 (human); retinitis pigmentosa 9 homolog; retinitis pigmentosa 9 protein; Retinitis pigmentosa 9 protein homolog; RGD1559759; RP9; RP9 pre-mRNA splicing factor; Rp9h
Gene Symbols RP9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RP9 (aa 172-221).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6100
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.