Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPA14 Monoclonal Antibody (11.1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA123279
Description
Recommended positive controls: MCF7, T24.
Replication protein A (RPA) is a single-stranded DNA binding protein. Human RPA is a heterotrimeric protein containing subunits of 70, 32 and 14 kDa. This protein complex is highly conserved in eukaryotes and is essential in DNA replication, homologous recombination and nucleotide excision repair. RPA has been shown to be a target for DNA-dependent protein kinase (DNA-PK) ataxia telangiectasia-mutated gene (ATM) and a cyclin dependent protein kinase.
Specifications
RPA14 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P35244, Q9CQ71 | |
Rpa3 | |
Full-length human RPA-14 expressed in E. coli.. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
11.1 | |
Unconjugated | |
Rpa3 | |
14kDa; C330026P08Rik; REPA3; Replication factor A protein 3; replication protein A 14 kDa subunit; replication protein A3; replication protein A3, 14kDa; RF-A protein 3; RP-A p14; RPA14; RPA3 | |
Mouse | |
Protein G | |
RUO | |
6119, 68240 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction