Learn More
Invitrogen™ RPA14 Polyclonal Antibody

Description
Immunogen sequence: DLPRSRINAG MLAQFIDKPV CFVGRLEKIH PTGKMFILSD GEGKNGTIEL MEPLDEEISG IVEVVGRVTA KATILCTSYV QFKEDSHPFD LGLYNEAVKI IHDFPQFYPL GIVQHD; Positive Samples: Mouse spleen; Cellular Location: Nucleus.
Specifications
Specifications
| Antigen | RPA14 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.14 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Rpa3 |
| Gene Accession No. | P35244, Q9CQ71 |
| Gene Alias | 14kDa; C330026P08Rik; REPA3; Replication factor A protein 3; replication protein A 14 kDa subunit; replication protein A3; replication protein A3, 14kDa; RF-A protein 3; RP-A p14; RPA14; RPA3 |
| Gene Symbols | Rpa3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.