Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPA2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100332PCP
Description
RPA2 Polyclonal antibody specifically detects RPA2 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
RPA2 | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
Maps to a region between residue 225 and the C-terminus (residue 270) of human Replication Protein A2 (32kD) using the numbering given in entry NP_002937.1 (GeneID 6118). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers | |
6118 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction