Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPA2, p Thr21 Antibody (3B2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | RPA2 |
---|---|
Clone | 3B2 |
Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
RPA2 | |
ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
A synthesized peptide derived from Human RPA2 [p Thr21] [UniProt P15927] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3B2 | |
ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
6118 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title