Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPA2, p Thr21 Antibody (SR1535), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321543100UL
Description
RPA2 Monoclonal antibody specifically detects RPA2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
RPA2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunoprecipitation | |
SR1535 | |
Western Blot 1:5000-1:10000, Immunoprecipitation 1:20 | |
REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34 | |
A synthesized peptide derived from human phospho-RPA2 [Thr21] (Uniprot #: P15927) | |
100 μg | |
Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers | |
6118 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction