Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582910 Supplier Invitrogen™ Supplier No. PA582910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HMVLSKANSQ PSAGRAPISN PGMSEAGNFG GNSFMPANGL TVAQNQVLNL IKACPRPEGL NFQDLKNQLK HMSVSSIKQA VDFLSNEGHI YSTVDDDH Highest antigen sequence indentity to the following orthologs: Rat - 82%, Mouse - 84%.

Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rpa2
Gene Accession No. P15927, Q62193, Q63528
Gene Alias 30-kDa protein; AA409079; AI325195; AU020965; HSSB; hypothetical protein; I79_004200; p32; p32-subunit of replication protein A; p34; REPA2; Replication factor A protein 2; replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2; replication protein A2, 32kDa; replication protein A4; RFA; RF-A protein 2; Rf-A2; RP21C; RP4-547C9.3; RP-A p32; RP-A p34; RPA2; RPA32; RPA34; RPA4; RpLP1; RpP2
Gene Symbols Rpa2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPA2. Recombinant protein control fragment (Product #RP-94131).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19891, 59102, 6118
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.