Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA2 Rabbit anti-Human, Unconjugated, Clone: RPA2/3140R, Abnova™

Catalog No. 89339600 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-600 Supplier Abnova Corporation Supplier No. RAB00621
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

RPA2 Monoclonal antibody specifically detects RPA2 in Human samples. It is validated for WB.

Specifications

Antigen RPA2
Applications Western Blot
Classification Monoclonal
Clone RPA2/3140R
Conjugate Unconjugated
Description replication protein A2, 32kDa
Gene RPA2
Gene Alias REPA2, RPA32
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human RPA2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6118
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.