Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575738 Supplier Invitrogen™ Supplier No. PA575738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The single-stranded-DNA-binding proteins (SSBs) are essential for DNA function in prokaryotic and eukaryotic cells, mitochondria, phages and viruses. Replication protein A (RPA), a highly conserved eukaryotic protein, is a heterotrimeric SSB consisting of three subunits that play an important role in DNA replication, recombination and repair. RPA is one of the major damage- recognition structures involved in the early stage of nucleotide excision repair and may function in telomere maintenance. The binding of human RPA (hRPA) to DNA involves molecular polarity, in which initial hRPA binding occurs on the 5' side of a ssDNA substrate and then extends in the 3' direction to create a stably bound hRPA. Widely expressed at low to intermediate levels, RPA 34 kDa subunit, which is also known as RPA4 (replication factor A protein 4), contains 261 amino acids, localizes to nucleus and is preferentially expressed in mucosa of colon and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RPA4
Gene Accession No. Q13156
Gene Alias HSU24186; MGC120333; MGC120334; Replication factor A protein 4; replication protein A 30 kDa subunit; replication protein A complex 34 kd subunit homolog Rpa4; replication protein A4; replication protein A4, 30kDa; replication protein A4, 34kDa; RF-A protein 4; RP-A p30; RPA4
Gene Symbols RPA4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human RPA4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29935
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.