Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPA4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5112927

Catalog No. PIPA5112927


Only null left
Add to Cart

Description

Description

RPA4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

The single-stranded-DNA-binding proteins (SSBs) are essential for DNA function in prokaryotic and eukaryotic cells, mitochondria, phages and viruses. Replication protein A (RPA), a highly conserved eukaryotic protein, is a heterotrimeric SSB consisting of three subunits that play an important role in DNA replication, recombination and repair. RPA is one of the major damage- recognition structures involved in the early stage of nucleotide excision repair and may function in telomere maintenance. The binding of human RPA (hRPA) to DNA involves molecular polarity, in which initial hRPA binding occurs on the 5' side of a ssDNA substrate and then extends in the 3' direction to create a stably bound hRPA. Widely expressed at low to intermediate levels, RPA 34 kDa subunit, which is also known as RPA4 (replication factor A protein 4), contains 261 amino acids, localizes to nucleus and is preferentially expressed in mucosa of colon and placenta.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPA4
Polyclonal
Unconjugated
RPA4
HSU24186; MGC120333; MGC120334; Replication factor A protein 4; replication protein A 30 kDa subunit; replication protein A complex 34 kd subunit homolog Rpa4; replication protein A4; replication protein A4, 30kDa; replication protein A4, 34kDa; RF-A protein 4; RP-A p30; RPA4
Rabbit
Antigen Affinity Chromatography
RUO
29935
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
0.56 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13156
RPA4
Recombinant Human Replication protein A 30 kDa subunit protein (1-230AA).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.