Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPA70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582269 Supplier Invitrogen™ Supplier No. PA582269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLSHTSGGTQ SKVVPIASLT PYQSKWTICA RVTNKSQIRT WSNSRGEGKL FSLELVDESG EIRATAFNEQ VDKFFPLIEV NKVYYFSKGT LKIANKQFTA VKNDYEMTFN NETSVMPCED DHHLPTVQFD FTGIDD Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 93%.

This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPA70
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPA1
Gene Accession No. P27694
Gene Alias 5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein
Gene Symbols RPA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPA1. Recombinant protein control fragment (Product #RP-89476).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6117
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.