Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPA70 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583821
Description
Immunogen sequence: PVLAIKGARV SDFGGRSLSV LSSSTIIANP DIPEAYKLRG WFDAEGQALD GVSISDLKSG GVGGSNTNWK TLYEVKSENL GQGDKPDYFS SVATVVYLRK ENCMYQACPT QDCNKKVIDQ QNGLYRCEK.
This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.
Specifications
RPA70 | |
Polyclonal | |
Unconjugated | |
RPA1 | |
5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6117 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P27694 | |
RPA1 | |
Recombinant protein corresponding to Human RPA1. Recombinant protein control fragment (Product #RP-104820). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction