Learn More
RPA70 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | RPA70 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2048
![]() |
Bethyl Laboratories, Inc
IHC00409T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2047
![]() |
Bethyl Laboratories, Inc
IHC00409 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rabbit, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Gray short-tailed
This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.Specifications
RPA70 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
MSTP075; RF-A protein 1; RP-A p70; replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; single-stranded DNA-binding protein | |
Between 350 and 400 | |
Primary | |
4° C | |
RPA1 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P27694, Q8VEE4 | |
6117, 68275 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.