Learn More
RPAIN Mouse anti-Human, Clone: OTI4F7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810499

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.Specifications
RPAIN | |
Monoclonal | |
Unconjugated | |
RPAIN | |
HRIP, RIP | |
Mouse | |
Affinity Chromatography | |
RUO | |
84268 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4F7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86UA6 | |
RPAIN | |
Full length human recombit protein of human RPAIN produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.