Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPAIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556770 Supplier Invitrogen™ Supplier No. PA556770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAESLRSPRR SLYKLVGSPP WKEAFRQRCL ERMRNSRDRL LNRYRQAGSS GPGNSQNSFL VQEVMEEEWN Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.

Replication protein A (RPA) is a single-stranded-DNA binding protein involved in numerous eukaryotic DNA processes including replication, repair and recombination. RPA interacting protein (RPA IP) has been identified as an adapter protein that is involved in RPA nuclear import instead of the prototypical importin proteins that normally mediate nuclear import. Multiple isoforms of RPA IP are known to exist, with the longest isoform localized to the cytoplasm. Isoform 2 is sumoylated and is located in the PML nuclear body within the nucleus. It has been suggested that this isoform mediates the localization of the RPA complex into the PML nuclear body, thereby participating in RPA function in DNA metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPAIN
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPAIN
Gene Accession No. Q86UA6
Gene Alias 2400006N03Rik; hRIP; nuclear transporter; RAP interaction protein; RAP interaction protein alpha; RGD1308492; RIP; RPA interacting protein; RPA interacting protein delta 2; RPA interacting protein epsilon; RPAIN; RPA-interacting protein
Gene Symbols RPAIN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPAIN. Recombinant protein control fragment (Product #RP-93237).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84268
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.