Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPAP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558334
Description
Immunogen sequence: SEKMPIEIEQ KPAQFATTVL PPIPANSFQL ESDFRQLKSS PDMLYQYLKQ IEPSLYPKLF QKNLDPDVFN QIVKILHDFY IEKEKPLLIF EILQRLSELK RFDMAVMFM Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
This gene encodes an RNA polymerase II-associated protein. The encoded protein may function in transcriptional regulation and may also regulate apoptosis. Alternatively spliced transcript variants have been described.
Specifications
RPAP3 | |
Polyclonal | |
Unconjugated | |
Rpap3 | |
2310042P20Rik; D15Ertd682e; RNA polymerase II associated protein 3; RNA polymerase II-associated protein 3; Rpap3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6T3 | |
Rpap3 | |
Recombinant protein corresponding to Human RPAP3. Recombinant protein control fragment (Product #RP-97623). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction