Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP253092
Description
RPB2 Polyclonal specifically detects RPB2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RPB2 | |
Polyclonal | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) | |
DNA-directed RNA polymerase II subunit RPB2, EC 2.7.7, EC 2.7.7.6, hRPB140, hsRPB2, POL2RB, polymerase (RNA) II (DNA directed) polypeptide B (140kD), polymerase (RNA) II (DNA directed) polypeptide B, 140kDa, RNA polymerase II subunit 2, RNA polymerase II subunit B2, RNA polymerase II subunit2, RNA polymerase II second largest subunit | |
Rabbit | |
Affinity Purified | |
RUO | |
5431 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7) with 0.01% Thimerosal | |
POLR2B | |
Recombinant protein encompassing a sequence within the N-terminus region of human RPB2. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction