Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPB8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP153016
Description
RPB8 Polyclonal specifically detects RPB8 in Human samples. It is validated for Western Blot.Specifications
RPB8 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
DNA-directed RNA polymerases I, II, and III subunit RPABC3, hsRPB8, II, and III 17.1 kDa polypeptide, II, and III subunit ABC3, polymerase (RNA) II (DNA directed) polypeptide H, RPB8 homolog | |
Rabbit | |
17 kDa | |
100 μL | |
Primary | |
This product is specific to Subunit or Isoform: RPABC3. | |
Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
P52434 | |
POLR2H | |
Synthetic peptides corresponding to POLR2H(polymerase (RNA) II (DNA directed) polypeptide H) The peptide sequence was selected from the N terminal of human POLR2H. Peptide sequence DLGDKFRLVIASTLYEDGTLDDGEYNPTDDRPSRADQFEYVMYGKVYRIE. | |
Protein A purified | |
RUO | |
5437 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction