Learn More
Description
Specifications
Specifications
| Antigen | RPE65 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, CyTOF |
| Classification | Monoclonal |
| Clone | 401.8B11.3D9 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 - 2 ug/ml, Simple Western, Flow Cytometry, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation reported by customer review, Immunohistochemistry-Paraffin 1:250-1:500, Immunohistochemistry-Frozen 1:250, In vivo assay reported in scientific literature (PMID 32173468), Immunofluorescence 1:50 - 1:200, CyTOF-ready, Knockout Validated |
| Gene Alias | All-trans-retinyl-palmitate hydrolase, EC 3.1.1.64, EC 5.2.1.7, LCA2p63, mRPE65, RBP-binding membrane protein, rd12, Retinal pigment epithelium-specific 65 kDa protein, retinal pigment epithelium-specific protein (65kD), retinal pigment epithelium-specific protein 65kDa, retinitis pigmentosa 20 (autosomal recessive), retinoid isomerohydrolase, Retinol isomerase, RP20, sRPE65 |
| Gene Symbols | RPE65 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
