Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPE65 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989175
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9175 Supplier Bioss Supplier No. BS9575R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPE65 is a major protein of the retinal pigment epithelium (RPE). RPE65 is essential for the regeneration of rhodopsin in the visual cycle. Mutations in RPE65 are responsible for certain forms of autosomal recessive severe retinal dystrophy, including Leber Congenital Amaurosis (LCA). Loss of function results in a block in retinoid processing required for the synthesis of 11-cis retinal and the accumulation of retinyl esters in the RPE.

Specifications

Antigen RPE65
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RPE65
Gene Accession No. Q16518
Gene Alias 65kDa; A930029L06Rik; All-trans-retinyl-palmitate hydrolase; BCO family, member 3; BCO3; BOS_3433; LCA2; Lutein isomerase; Meso-zeaxanthin isomerase; modifier of retinal degeneration 1; Mord1; mRPE65; p63; RBP-binding membrane protein; rd12; retinal pigment epithelium 65; retinal pigment epithelium 65-like; retinal pigment epithelium abundant protein RPE65; retinal pigment epithelium specific protein 65; retinal pigment epithelium, 65 kDa; Retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific protein (65kD); retinal pigment epithelium-specific protein 65kDa; Retinitis pigmentos; retinitis pigmentosa 20 (autosomal recessive); Retinoid isomerohydrolase; Retinol isomerase; RP20; Rpe65; rpe65 gene for retinal pigment epithelium-specific protein; RPE65, retinoid isomerohydrolase; sRPE65; truncated RPE65
Gene Symbols RPE65
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RPE65.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6121
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.