Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPE65 Recombinant Rabbit Monoclonal Antibody (7B10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549847
Description
RPE65 Recombinant Monoclonal Antibody for IHC (P), ELISA
RPE65 is a major protein of the retinal pigment epithelium (RPE). RPE65 is essential for the regeneration of rhodopsin in the visual cycle. Mutations in RPE65 are responsible for certain forms of autosomal recessive severe retinal dystrophy, including Leber Congenital Amaurosis (LCA). Loss of function results in a block in retinoid processing required for the synthesis of 11-cis retinal and the accumulation of retinyl esters in the RPE.
Specifications
RPE65 | |
Recombinant Monoclonal | |
1.6 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q16518 | |
RPE65 | |
A synthesized peptide derived from human RPE65. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
7B10 | |
Unconjugated | |
RPE65 | |
65kDa; A930029L06Rik; all-trans-retinyl-palmitate hydrolase; BCO family, member 3; BCO3; LCA2; Lutein isomerase; membrane receptor p63; meso-zeaxanthin isomerase; modifier of retinal degeneration 1; Mord1; mRPE65; p63; RBP-binding membrane protein; rd12; retinal pigment epithelium 65; retinal pigment epithelium 65-like; retinal pigment epithelium abundant protein RPE65; retinal pigment epithelium specific protein 65; retinal pigment epithelium, 65 kDa; retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific 65kD protein; retinal pigment epithelium-specific protein (65kD); retinal pigment epithelium-specific protein 65kDa; retinal pigment epithelium-specific protein RPE65; Retinitis pigmentos; retinitis pigmentosa 20 (autosomal recessive); retinoid isomerohydrolase; retinoid isomerohydrolase RPE65; retinol isomerase; RP20; RPE65; rpe65 gene for retinal pigment epithelium-specific protein; RPE65, retinoid isomerohydrolase; sRPE65; truncated RPE65 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6121 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction