Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPEL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524517
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524517 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524517 Supplier Invitrogen™ Supplier No. PA524517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPEL1 Polyclonal Antibody for Western Blot, Flow

Catalyzes the reversible epimerization of D-ribulose 5- phosphate to D-xylulose 5-phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPEL1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RPEL1
Gene Accession No. Q2QD12
Gene Alias 9630030F18Rik; dJ257A7.2; hCG_2024410; hCG2024410; KIAA1733; PHACTR1; Phosphatase and actin regulator 1; rcRPE protein; ribulose-5-phosphate-3-epimerase like 1; ribulose-5-phosphate-3-epimerase-like 1; ribulose-5-phosphate-3-epimerase-like protein 1; Ribulose-phosphate 3-epimerase-like protein 1; RPEL; RPEL repeat containing 1; RPEL1
Gene Symbols RPEL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 187-215 amino acids from the C-terminal region of human RPEL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729020
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.