Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPGR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551607 Supplier Invitrogen™ Supplier No. PA551607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EINDTCLSVA TFLPYSSLTS GNVLQRTLSA RMRRRERERS PDSFSMRRTL PPIEGTLGLS ACFLPNSVFP RCSERNLQES VLSEQDLMQP EEPDYLLDEM TKEAEIDNSS TVESLGETTD ILNMTHIMSL N Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 41%.

This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPGR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPGR
Gene Accession No. Q92834
Gene Alias COD1; CORDX1; CRD; mRpgr; orf15; PCDX; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; retinitis pigmentosa GTPase regulator; RP15; RP3; Rp3h; Rpgr; X-linked retinitis pigmentosa GTPase regulator; XLRP3
Gene Symbols RPGR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPGR. Recombinant protein control fragment (Product #RP-92146).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6103
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.