Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPGR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA567290
Description
Immunogen sequence: PFFGNLPDRGM NTESEENKDF VKKRESCKQD VIFDSERESV EKPDSYMEGA SESQQGIADG FQQPEAIEFS SGEKEDDEVE TDQNIRYGRK LIEQG Highest antigen sequence identity to the following orthologs - mouse 48%, rat 29%.
This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
Specifications
RPGR | |
Polyclonal | |
Unconjugated | |
RPGR | |
X-linked retinitis pigmentosa GTPase regulator; RPGR; RP3; XLRP3; CRD; COD1; PCDX; RP15; orf15; CORDX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6103 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92834 | |
RPGR | |
Recombit Human RPGR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction