Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPGRIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560172 Supplier Invitrogen™ Supplier No. PA560172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASPEVPIEA GQYRSKRKPP HGGERKEKEH QVVSYSRRKH GKRIGVQGKN RMEYLSLNIL NGNTPEQVNY TEWKFSETNS FIGDGFKNQH EEEEMTLSHS ALKQKEPLHP VNDKESSEQG SEVSEAQTTD SDDVIVPPMS QKYPKADSEK Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 23%.

May function as scaffolding protein. Required for normal location of RPGR at the connecting cilium of photoreceptor cells. Required for normal disk morphogenesis and disk organization in the outer segment of photoreceptor cells and for survival of photoreceptor cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPGRIP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPGRIP1
Gene Accession No. Q96KN7
Gene Alias 0610005A07Rik; 4930401L23Rik; 4930505G06Rik; A930002K18Rik; AA415034; CORD13; LCA6; nmf247; retinitis pigmentosa GTPase regulator interacting protein 1; RGI1; RGRIP; RPGR-interacting protein 1; RPGRIP; Rpgrip1; RPGRIP1d; X-linked retinitis pigmentosa GTPase regulator-interacting protein 1
Gene Symbols RPGRIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPGRIP1. Recombinant protein control fragment (Product #RP-89028).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57096
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.