Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPGRIP1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728578 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8578 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8578 Supplier Proteintech Group Inc Supplier No. 132141AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a photoreceptor protein that interacts with retinitis pigmentosa GTPase regulator protein and is a key component of cone and rod photoreceptor cells. Mutations in this gene lead to autosomal recessive congenital blindness.

Specifications

Antigen RPGRIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPGRIP1
Gene Accession No. Q96KN7
Gene Alias CORD9, LCA6, RGI1, RGRIP, RPGR interacting protein 1, RPGRIP, RPGRIP1, RPGRIP1d
Gene Symbols RPGRIP1
Host Species Rabbit
Immunogen RPGRIP1 Fusion Protein Ag4089
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57096
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.