Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPGRIP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565271 Supplier Invitrogen™ Supplier No. PA565271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKYGNSLLEEA RGEIRNLENV IQSQRGQIEE LEHLAEILKT QLRRKENEIE LSLLQLREQQ ATDQRSNIRD NVEMIKLHKQ LVEKSNALSA MEGKFIQLQE KQRTLRIS Highest antigen sequence identity to the following orthologs - mouse 95%, rat 94%.

The protein encoded by this gene can localize to the basal body-centrosome complex or to primary cilia and centrosomes in ciliated cells. The encoded protein has been found to interact with nephrocystin-4. Defects in this gene are a cause of Joubert syndrome type 7 and Meckel syndrome type 5. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPGRIP1L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPGRIP1L
Gene Accession No. Q68CZ1
Gene Alias 1700047E16Rik; 4931437C01; CORS3; DKFZp686C0668; fantom; fantom homolog; Ftm; JBTS7; KIAA1005; MKS5; Nephrocystin-8; NPHP8; PPP1R134; Protein fantom; protein phosphatase 1, regulatory subunit 134; RGD1311099; RPGR-interacting protein 1-like protein; RPGRIP1 like; RPGRIP1L; Rpgrip1-like; RPGRIP1-like protein
Gene Symbols RPGRIP1L
Host Species Rabbit
Immunogen Recombinant Human RPGRIP1L. Recombinant protein control fragment (Product #RP-105989).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.