Learn More
Invitrogen™ RPH3A Polyclonal Antibody

Description
Immunogen sequence: SPAGLRRANS VQASRPAPGS VQSPAPPQPG QPGTPGGSRP GPGPAGRFPD QKPEVAPSDP GTTAPPREER TGGVGGYPAV GAREDRMSHP SGPYSQASAA APQPAAARQP PPPEEEEEEA NSYDSDEATT LGALEFSLLY DQDNS Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.
Specifications
Specifications
| Antigen | RPH3A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RPH3A |
| Gene Accession No. | Q9Y2J0 |
| Gene Alias | 2900002P20Rik; AU022689; AW108370; Exophilin 1; Exophilin1; Exophilin-1; KIAA0985; rabphilin 3A; rabphilin 3A homolog; rabphilin-3A; RPH3A |
| Gene Symbols | RPH3A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.