Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPH3AL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546737 Supplier Invitrogen™ Supplier No. PA546737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RIYTWARGRV VSSDSDSDSD LSSSSLEDRL PSTGVRDRKG DKPWKESGGS Sequence homology: Cow: 79%; Human: 100%.

The protein encoded by this gene plays a direct regulatory role in calcium-ion-dependent exocytosis in both endocrine and exocrine cells and plays a key role in insulin secretion by pancreatic cells. This gene is likely a tumor suppressor. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPH3AL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RPH3AL
Gene Accession No. Q9UNE2
Gene Alias Gm1753; no C2 domains protein; NOC2; rab effector Noc2; rabphilin 3A like (without C2 domains); rabphilin 3A-like (without C2 domains); rabphilin-3A-like protein; Rph3al
Gene Symbols RPH3AL
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human RPH3AL (aa 196-245).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9501
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.