Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPIA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560051
Description
Immunogen sequence: AVENHVRNNQ VLGIGSGSTI VHAVQRIAER VKQENLNLVC IPTSFQARQL ILQYGLTLSD LDRHPEIDLA ID Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
Defects in RPIA are the cause of ribose 5-phosphate isomerase deficiency. The exact function of RPIA remains unknown.
Specifications
RPIA | |
Polyclonal | |
Unconjugated | |
RPIA | |
Phosphoriboisomerase; ribose 5-phosphate epimerase; ribose 5-phosphate isomerase A; ribose 5-phosphate isomerase A (ribose 5-phosphate epimerase); ribose-5-phosphate isomerase; RPI; Rpia; RPIAD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49247 | |
RPIA | |
Recombinant protein corresponding to Human RPIA. Recombinant protein control fragment (Product #RP-96149). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction