Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPIP8 Antibody (OTI3C6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273943
Description
RPIP8 Monoclonal specifically detects RPIP8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RPIP8 | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000, Immunohistochemistry 1:150 | |
Rap2-interacting protein 8, RAP2IPRUN domain-containing protein 3A, RPIP-8, RPIP8RaP2 interacting protein 8, RUN domain containing 3A | |
Mouse | |
45.5 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3C6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
RUNDC3A | |
Full length human recombinant protein of human RUNDC3A(NP_006686) produced in HEK293T cell. | |
Affinity Purified | |
Signal Transduction | |
10900 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction