Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL10 Monoclonal Antibody (OTI9A8)
GREENER_CHOICE

Catalog No. PIMA526904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526904 Supplier Invitrogen™ Supplier No. MA526904
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPL10 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a ribosomal protein that is a component of the 60S ribosome subunit. The related protein in chicken can bind to c-Jun and can repress c-Jun-mediated transcriptional activation. Some studies have detected an association between variation in this gene and autism spectrum disorders, though others do not detect this relationship. There are multiple pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI9A8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rpl10
Gene Accession No. P27635
Gene Alias 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; NOV; Protein QM; Protein QM homolog; QM; QM16; ribosomal protein 10; ribosomal protein L10; Rpl10; RPLPO; Tumor suppressor QM; Wilms tumor-related protein
Gene Symbols Rpl10
Host Species Mouse
Immunogen Full length human recombinant protein of RPL10 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6134
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.