Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552816
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552816 Supplier Invitrogen™ Supplier No. PA552816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KMSSCAGADR LQTGMRGAFG KPQGTVARVH IGQVIMSIRT KLQNKEHVIE ALRRAKFKFP GRQKIHISKK WGFTKFNADE FEDMVAEKRL IPDGCGVKYI PSRGPLDKWW Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gene encodes a ribosomal protein that is a component of the 60S ribosome subunit. The related protein in chicken can bind to c-Jun and can repress c-Jun-mediated transcriptional activation. Some studies have detected an association between variation in this gene and autism spectrum disorders, though others do not detect this relationship. There are multiple pseudogenes of this gene dispersed throughout the genome. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL10
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rpl10
Gene Accession No. P27635, Q6PDV7, Q6ZWV3
Gene Alias 24.6 kDa protein; 24.6kDa protein; 60S ribosomal protein L10; AUTSX5; D0HXS648; DXHXS648; DXHXS648E; DXS648; DXS648E; L10; Laminin receptor homolog; Large ribosomal subunit protein uL16; NOV; Protein QM; Protein QM homolog; QM; QM16; ribosomal protein 10; ribosomal protein L10; Rpl10; RPLPO; Tumor suppressor QM; Wilms tumor-related protein
Gene Symbols Rpl10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL10. Recombinant protein control fragment (Product #RP-102219).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110954, 6134, 81764
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.