Learn More
Invitrogen™ RPL10L Polyclonal Antibody

Description
The antibody detects endogenous levels of total RPL10L protein.
Specifications
Specifications
| Antigen | RPL10L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RPL10L |
| Gene Accession No. | Q96L21 |
| Gene Alias | 60S ribosomal protein L10-like; EG238217; Large ribosomal subunit protein uL16-like; ribosomal protein L10 like; ribosomal protein L10-like; RL10L; RPL10_5_1358; Rpl10l |
| Gene Symbols | RPL10L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.