Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL10L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598749
Description
RPL10L Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a protein sharing sequence similarity with ribosomal protein L10. It is not currently known whether the encoded protein is a functional ribosomal protein or whether it has evolved a function that is independent of the ribosome. This gene is intronless. [provided by RefSeq, Jul 2008].
Specifications
RPL10L | |
Polyclonal | |
Unconjugated | |
RPL10L | |
60S ribosomal protein L10-like; EG238217; Large ribosomal subunit protein uL16-like; ribosomal protein L10 like; ribosomal protein L10-like; RL10L; RPL10_5_1358; Rpl10l | |
Rabbit | |
Protein G | |
RUO | |
140801, 238217 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P86048, Q96L21 | |
RPL10L | |
Recombinant Human 60S ribosomal protein L10-like protein (1-214AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction