Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584963 Supplier Invitrogen™ Supplier No. PA584963
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQDQGEKEN PMRELRIRKL CLNICVGESG DRLTR Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

Subunit of the splicing factor SF3B required for 'A' complex assembly formed by the stable binding of U2 snRNP to the branchpoint sequence (BPS) in pre-mRNA. Sequence independent binding of SF3A/SF3B complex upstream of the branch site is essential, it may anchor U2 snRNP to the pre-mRNA. May also be involved in the assembly of the 'E' complex. Belongs also to the minor U12-dependent spliceosome, which is involved in the splicing of rare class of nuclear pre-mRNA intron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL11
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rpl11
Gene Accession No. P62913
Gene Alias 2010203J19Rik; 60S ribosomal protein L11; cell growth-inhibiting protein 34; CLL-associated antigen KW-12; DBA7; GIG34; L11; Large ribosomal subunit protein uL5; ribosomal protein L11; RP11-223J15.3; RPL11; zgc:86748
Gene Symbols Rpl11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPL11. Recombinant protein control fragment (Product #RP-108021).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.