Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPL12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551048
Description
RPL12 Polyclonal Antibody detects endogenous levels of RPL12 protein.
Eukaryotic translation initiation factor-5.
Specifications
RPL12 | |
Polyclonal | |
Unconjugated | |
RPL12 | |
60S ribosomal protein L12; E430018F03; L12; Large ribosomal subunit protein uL11; ribosomal protein L12; Rpl12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
269261, 499782, 6136 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P23358, P30050, P35979 | |
RPL12 | |
Synthesized peptide derived from the Internal region of human Ribosomal Protein L12 (aa 61-110). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction