Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157476
Description
RPL13 Polyclonal specifically detects RPL13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RPL13 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
BBC1FLJ27454, Breast basic conserved protein 1, D16S444E, FLJ27453, MGC117342, MGC71373,60S ribosomal protein L13, OK/SW-cl.46, ribosomal protein L13 | |
Rabbit | |
24 kDa | |
100 μL | |
Core ESC Like Genes, Stem Cell Markers | |
6137 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P26373 | |
RPL13 | |
Synthetic peptides corresponding to RPL13 (ribosomal protein L13) The peptide sequence was selected from the C terminal of RPL13 (NP_000968). Peptide sequence KKEKARVITEEEKNFKAFASLRMARANARLFGIRAKRAKEAAEQDVEKKK. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Xenopus: 92%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction