Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL13A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517176 Supplier Invitrogen™ Supplier No. PA517176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L13P family of ribosomal proteins. It is located in the cytoplasm. Transcript variants utilizing alternative polyA signals have been observed. This gene is co-transcribed with the small nucleolar RNA genes U32, U33, U34, and U35, which are located in its second, fourth, fifth, and sixth introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL13A
Applications Western Blot
Classification Polyclonal
Concentration 58 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene RPL13A
Gene Accession No. P40429
Gene Alias 1810026N22Rik; 23 kDa highly basic protein; 60S ribosomal protein L13a; L13A; Large ribosomal subunit protein uL13; P198; ribosomal protein L13a; Rpl13a; tissue specific transplantation antigen 1; tissue specific transplantation antigen P198-7; transplantation antigen P198; TSTA1; Tstap198-7; tum-antigen; Tum-P198 antigen; tum-transplantation antigen P198
Gene Symbols RPL13A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence of human ribosomal protein L13a.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23521
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.