Learn More
Invitrogen™ RPL13A Polyclonal Antibody

Description
Predicted to react with bovine, human, monkey, pig and rat based on sequence homology.
Specifications
Specifications
| Antigen | RPL13A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RPL13A |
| Gene Accession No. | P19253 |
| Gene Alias | 1810026N22Rik; 23 kDa highly basic protein; 60S ribosomal protein L13a; L13A; Large ribosomal subunit protein uL13; P198; ribosomal protein L13a; Rpl13a; tissue specific transplantation antigen 1; tissue specific transplantation antigen P198-7; transplantation antigen P198; TSTA1; Tstap198-7; tum-antigen; Tum-P198 antigen; tum-transplantation antigen P198 |
| Gene Symbols | RPL13A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.