Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPL14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118895 Supplier Invitrogen™ Supplier No. PA5118895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RPL14.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L14E family of ribosomal proteins. It contains a basic region-leucine zipper (bZIP)-like domain. The protein is located in the cytoplasm. This gene contains a trinucleotide (GCT) repeat tract whose length is highly polymorphic; these triplet repeats result in a stretch of alanine residues in the encoded protein. Transcript variants utilizing alternative polyA signals and alternative 5'-terminal exons exist but all encode the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Aoki M., Diabetes 45:157-164(1996). Tanaka M., Biochem. Biophys. Res. Commun. 243:531-537(1998). Yoshihama M., Genome Res. 12:379-390(2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPL14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPL14
Gene Accession No. P50914, Q63507, Q9CR57
Gene Alias 3100001N19Rik; 60S ribosomal protein L14; AA407502; AL022816; CAG-ISL 7; CAG-ISL-7; CTG-B33; hRL14; L14; Large ribosomal subunit protein eL14; MGC88594; ribosomal protein L14; RL14; RPL14
Gene Symbols RPL14
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPL14(Accession P50914), corresponding to amino acid residues I82-K132.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65043, 67115, 9045
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.